2-Methylbutaan ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

270342-1L 270342-2.5L
270342-1LEA 228 EUR
270342-1L 270342-2.5L
2-Methylbutaan ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
2-Methylbutaan

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formule: CH₃CH₂CH(CH₃)₂
Kookpunt: 28 °C (1,013 hPa)
Smeltpunt: –160 °C
Dichtheid: 0,62 g/cm³ (20 °C)
Vlampunt: <–35 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009338
CAS nummer: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 240 nm Max. 0.01
Absorbance at 220 nm Max. 0.07
Absorbance at 210 nm Max. 0.30
Absorbance at 192 nm Max. 1.0
Vapor Density 2.6 (vs air)
Autoignition Temperature 788 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR