Potassium thiocyanate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Potassium rhodanide

27035.230EA 52.3 EUR
27035.230 27035.460
Potassium thiocyanate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium thiocyanate
Formule: KSCN
Kookpunt: 500 °C (1013 hPa)
Smeltpunt: 175 °C
Dichtheid: 1,61 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011413
CAS nummer: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (25°C; 5 %) 5.3 - 8.5
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 30 ppm
Substances reducing iodine (as I) Max. 0.025 %
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 20 ppm
S (Sulphide) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
Cu (Copper) Max. 2 ppm
Fe (Iron) Max. 1 ppm
Na (Sodium) Max. 50 ppm
Pb (Lead) Max. 2 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR