di-Sodium tetraborate, watervrij ≥98%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

27716.290EA 101 EUR
27716.290 27716.368
di-Sodium tetraborate, watervrij ≥98%, GPR RECTAPUR®
di-Sodium tetraborate
Formule: Na₂B₄O₇
Kookpunt: 1575 °C (1013 hPa)
Smeltpunt: 742 °C
Dichtheid: 2,38 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081185
CAS nummer: 1330-43-4
EINECS: 215-540-4
Bereik: 01-2119490790-32
Merck Index: 12,08733

Order Now

Specification Test Results

Assay Min. 98 %
Water Max. 1.0%
Sulphate (SO4) Max. 150 ppm
Fe (Iron) Max. 40 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR