Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Synoniemen: Sodium bicarbonate
27776.296EA 29.4 EUR
27776.296 27776.365 27776.460
Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Sodium hydrogen carbonate
Formule: NaHCO₃
MW: 84.01 g/mol
Smeltpunt: 270
Dichtheid: 2.159 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003528
CAS nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 150 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm