Natriumchloride 98, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Saline, NaCl, Sea salt, Halite

27788.297EA 14.2 EUR
27788.297 27788.366 27788.460
Natriumchloride 98, TECHNICAL
Natriumchloride
Formule: NaCl
MW: 58.44 g/mol
Kookpunt: 1413
Smeltpunt: 801
Dichtheid: 2.16 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay Min. 98 %
Appearance Colourless crystals


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR