tri-Sodium citrate dihydrate 99.0-100.5% (dry basis), crystallised Ph. Eur., BP, USP

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

27831.297EA 84.1 EUR
27831.297 27831.366 27831.468
tri-Sodium citrate dihydrate 99.0-100.5% (dry basis), crystallised Ph. Eur., BP, USP
tri-Sodium citrate dihydrate

Appearance: White or almost white, crystalline powder or white or almost white, granular crystals, slightly deliquescent in moist air.


Opgelet: Not for use as Active Pharmaceutical Ingredient (API).

Formule: Na₃C₆H₅O₇·2H₂O
Smeltpunt: 150 °C
Dichtheid: 1,76 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150031
CAS nummer: 6132-04-3
EINECS: 200-675-3
Bereik: 01-2119457027-40
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) Max. 10 ppm
Residual solvents Passes test
As (Arsenic) Max. 1 ppm
Cd (Cadmium) Max. 1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 2 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 3 ppm
V (Vanadium) Max. 2 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: .
Assay (on anhydrous) [Ph.Eur./BP] 99,0 --> 101,0 %
Identification A Passes test USP
Identification B Passes test USP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
Readily carbonisable substances Passes test Ph.Eur./BP
Cl (Chloride) [Ph.Eur./BP] Max. 50 ppm
C2O4 (Oxalate) [Ph.Eur./BP] Max. 300 ppm
SO4 (Sulphate) [Ph.Eur./BP] Max. 150 ppm
Water [Ph.Eur./BP] 11,0 --> 13,0 %
USP specifications: .
Assay (calculated on anhydrous) [USP] 99,0 --> 100,5 %
Identification C Passes test USP
Tartrate Passes test USP
Alkalinity Passes test USP
Water determination [USP] 10,0 --> 13,0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR