Dinatriumdisulfiet ≥97%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

27921.298EA 38.1 EUR
27921.298 27921.367
Dinatriumdisulfiet ≥97%, GPR RECTAPUR®
Dinatriumdisulfiet
Formule: Na₂S₂O₅
Smeltpunt: 150 °C
Dichtheid: 1,48 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167602
CAS nummer: 7681-57-4
EINECS: 231-673-0
Bereik: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 97 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 0.02 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR