Ion exchange chromatography column, HiScreen™ Capto™ MMC

Leverancier: Cytiva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

28-9269-80
28-9269-80EA 170 EUR
28-9269-80
Ion exchange chromatography column, HiScreen™ Capto™ MMC
Kolommen voor chromatografie
HiScreen Capto MMC columns are prepacked with Capto MMC ion exchange chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR