Affinity chromatography column, HiScreen™ IMAC FF
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
28-9505-17
28-9505-17EA 197 EUR
28-9505-17
Affinity chromatography column, HiScreen™ IMAC FF
Kolommen voor chromatografie
HiScreen™ IMAC FF columns for optimising purification of histidine-tagged proteins by Immobilised Metal ion Affinity Chromatography (IMAC).
  • Conveniently charge with metal ion of choice (Cu²⁺, Zn²⁺, Ni²⁺ or Co²⁺)
  • Prepacked HiScreen™ columns for method optimization and process development
  • High binding capacity
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
  • Flow velocity: <600 cm/h; The pressure over the packed bed varies depending on a range of parameters such as the characteristics of
More Product Information