Sodium sulphite 95.0-100.5%, purified

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

28126.297EA 26 EUR
28126.297 28126.366 28126.460
Sodium sulphite 95.0-100.5%, purified
Sodium sulphite
Formule: Na₂SO₃
MW: 126.04 g/mol
Smeltpunt: > 500 °C
Dichtheid: 2.63 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003503
CAS nummer: 7757-83-7
EINECS: 231-821-4
Bereik: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Assay (at filling) 95.0 - 100.5 %
Heavy metals (as Pb) Max. 50 ppm
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR