Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate

28195.238EA 136 EUR
28195.238 28195.460
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Sodium tungstate dihydrate

This product is not suitable for the Colin reaction.

Formule: Na₂WO₄·2H₂O
Smeltpunt: 100 °C
Dichtheid: 3,23 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149190
CAS nummer: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847

Order Now

Specification Test Results

Assay Min. 99 %
Cl (Chloride) Max. 30 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 5 ppm
Mo (Molybdenum) Max. 50 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR