Thioaceetamide

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ethanethioamide

28600.183EA 0 EUR
28600.183
Thioaceetamide
Thioaceetamide
Formule: CH₃CSNH₂
MW: 75.13 g/mol
Smeltpunt: 113…114 °C
Dichtheid: 1.336 g/cm³ (20 °C)
MDL Nummer: MFCD00008070
CAS nummer: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR