Tributylfosfaat ≥99.0%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

28726.291EA 57.7 EUR
28726.291 28726.462
Tributylfosfaat ≥99.0%, GPR RECTAPUR®
Tributylfosfaat
Formule: (CH₃(CH₂)₃O)₃PO
Kookpunt: 289 °C (1013 hPa)
Smeltpunt: –79 °C
Dichtheid: 0,982 g/cm³ (20 °C)
Vlampunt: 151 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009436
CAS nummer: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99,0 %
Appearance Clear liquid
Colouration Max. 50 APHA
Density (20/4) 0,975 --> 0,980
Refractive index (20°C) 1,423 --> 1,425
Water Max. 0,2 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR