2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

28780.322EA 125 EUR
28780.322 28780.366
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
2,2,4-Trimethylpentane
Formule: (CH₃)₂CHCH₂C(CH₃)₃
Kookpunt: 99,2 °C (1013 hPa)
Smeltpunt: –107 °C
Dichtheid: 0,692 g/cm³ (20 °C)
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
Density (20/4) 0.690 - 0.695
Free acidity Max. 0.0004 meq/g
Evaporation residue Max. 50 ppm
Water Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR