Small-scale SEC columns, Superose 6 Increase
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29-0915-96 29-0915-97 29-0915-98
29-0915-96EA 2900 EUR
29-0915-96 29-0915-97 29-0915-98
Small-scale SEC columns, Superose 6 Increase
Kolommen voor chromatografie
Superose 6 Increase SEC columns are designed for high-resolution, small-scale preparative purification, and analysis of large globular proteins, protein complexes, and other macromolecules in the molecular weight range 5 to 5000 kD.
  • Wide fractionation range: Optimized to purify large biomolecules, including membrane proteins, protein complexes, and DNA fragments
  • Very high resolution: Small bead size and a narrow particle size distribution provide high resolution, for high protein purity
  • High flow rates: Rigid beads give excellent pressure/flow properties