Potassium acetate 99,0, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid potassium salt

2912-REA 2700.45 EUR
2912-R 2912-05
Potassium acetate 99,0, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium acetate
Formule: CH₃COOK
MW: 98.14 g/mol
Smeltpunt: 304
Dichtheid: 1.57 g/cm³ (25 °C)
Vlampunt: >250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012458
CAS nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements = 99.0 %
For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (CH₃COOK) (by non-aqueous titration) ≥99.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.5 - 9.0
Chloride (Cl) ≤0.003 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.03 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR