Zinkoxide light ≥98%, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Zinci oxidum, Zinc (II) oxide

29199.295EA 54.9 EUR
29199.295
Zinkoxide light ≥98%, TECHNICAL
Zinkoxide
Formule: ZnO
Kookpunt: 2360 °C (1013 hPa)
Smeltpunt: 1975 °C
Dichtheid: 5,6 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
Merck Index: 12,10279

Order Now

Specification Test Results

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR