Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

29247.292EA 51.9 EUR
29247.292 29247.361
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
Zinc sulphate heptahydrate
Formule: ZnSO₄·7H₂O
MW: 287.54 g/mol
Smeltpunt: 100
Dichtheid: 1.97 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149894
CAS nummer: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9 III
Merck Index: 12,10293

Order Now

Specification Test Results

Assay Min. 99 %
Cl (Chloride) Max. 0.03 %
Fe (Iron) Max. 20 ppm
Pb (Lead) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR