Dubbelpak hersluitbare sluitingen SJ354D, Dual Lock™
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SJ354B SJ354D
293-0005EA 45.2 EUR
293-0005 293-0014
Dubbelpak hersluitbare sluitingen SJ354D, Dual Lock™
Kleefstoffen
This reclosable fastener has a rubber based adhesive on the back of a black, polypropylene reclosable fastener. It's suitable for bonding many plastics, powder coated paints, and other low surface energy materials.
  • Snap-in-place installation replaces screws, nails, snaps, and rivets
  • Type 250 mates to Type 170, 250, or 400
  • Closes with an audible snap
  • Opens and closes 1000 times before losing 50% of original tensile strength
  • Pressure-sensitive adhesive provides excellent initial tack to most surfaces
  • Helps reduce vibration
Designed for use with manyMore Product Information
New Product
Order Now
Specificaties
Environmentally Preferable