Trifluorazijnzuur ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: TFA

299537-100G 299537-500G 299537-25G
299537-100GEA 236 EUR
299537-100G 299537-500G 299537-25G
Trifluorazijnzuur ≥99%, BioSyn™ for protein sequencing, purified by redistillation, Fluka™
Trifluorazijnzuur
Formule: C₂HF₃O₂
Kookpunt: 72 °C (1,013 hPa)
Smeltpunt: -15 °C
Dichtheid: 1,48 g/cm³ (20 °C)
Vlampunt: 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004169
CAS nummer: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Assay 99.0 - 101.0 %
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.05 %
Suitability complying
APHA Max. 25


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR