Anti-ITGAM Rat Monoclonal Antibody (Biotin) [clone: M1/70]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30-0112-U500 30-0112-U100 30-0112-U025
30-0112-U500EA 102 EUR
30-0112-U500 30-0112-U100 30-0112-U025
Anti-ITGAM Rat Monoclonal Antibody (Biotin) [clone: M1/70]
Antilichamen
The M1/70 antibody reacts with human and mouse CD11b, also known as integrin alpha M. This 165-170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles withMore Product Information