Forceps, disposable
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5378-1041 5378-0041 5378-1042 5378-0042 5378-1040 5378-0040
300-0346EA 61.6 EUR
300-0346 300-0347 300-0348 300-0349 300-0344 300-0345
Forceps, disposable
Pincetten
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
  • PS, white
  • Can be individually packed and sterilised
  • For single use
  • Clean room manufactured (Class 7 cleanroom)
  • Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. Light plastic forceps for single use allow for fatigue-free work. Cleanroom manufactured lab assistants meet the highest standards and prevent cross-contamination orMore Product Information