Polyurethane surface sampling bags with PolyProbe™, Whirl-Pak®

Leverancier: NASCO FORT ATKINSON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B01592WA B01613
300-0674EA 626 EUR
300-0674 NASCB01613
Polyurethane surface sampling bags with PolyProbe™, Whirl-Pak®
Zakken Monsterzakken
Make environmental surface sampling faster and easier with sterile Whirl-Pak polyurethane surface sampling bags with 8" PolyProbe™.

  • Write-on label makes identifying samples easy and convenient
  • Available with dry PolyProbe or hydrated with 10 ml of HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility (+1 Sterilization)
  • PolyProbe comes with a probe attached proven to be very strong – holds up to pressures required to breakthrough biofilms
  • Opening Tyvek® Pull Tabs are perpendicular to the opening of the bag: Keeps your hands away for a
More Product Information
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR