Serum substitute, BaseSerum™

Leverancier: SEQENS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

30001C-100ML 30001C-500ML 30001C-1L
30001C-100MLEA 155 EUR
30001C-100ML 30001C-500ML 30001C-1L
Serum substitute, BaseSerum™
Serum
SEQENS IVD BaseSerum™ is a human serum substitute obtained from human plasma.

  • Total proteins: 60 to 80 g/L
  • Fibrinogen: Negative
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

It is electrolytes balanced to match physiological conditions. For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Opgelet: Only for research orMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR