Zoutzuur 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetrische oplossing

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

30024.324EA 54.7 EUR
30024.324 30024.370 30024.404 30024.415 30024.448 30024.290
Zoutzuur 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetrische oplossing
Zoutzuur
Formule: HCl
Dichtheid: 1,02 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Conforms to Reag. Ph.Eur. Passes test
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
Conforms to Reag. USP-NF Passes test
NIST traceable Confirmed

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR