Protein quantitation kit, AccuOrange™
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30071 30071-T
30071.EA 396 EUR
30071. 30071-T
Protein quantitation kit, AccuOrange™
Eiwitextractie Protein Extraction Kits
AccuOrange™ Protein Quantitation Kit is a highly sensitive fluorescence-based assay for quantitating purified protein samples in 96-well format (excitation/emission = 480/598 nm).
  • Quantitating purified protein samples in 96-well format
  • More sensitive than traditional protein quantitation assays
  • Superior linearity and reproducibility
  • Compatible with the AccuLite™ 470 Mini fluorometer
  • Shows minimal variability between different proteins, and has stable fluorescence signal for up to 16 hours
  • Detection range: 0,1 to 15 µg/ml protein
AccuOrange™ is recommended for quantitating purified protein or antibody samples. The AccuOrange™ assay has low tolerance for non-ionic detergents, and is not recommended for use with cell lysates containing Triton X-100, sodium deoxycholate, CHAPs, or other non-ionic detergents. The assay can tolerate up to 0,01% SDS (final concentration in assay)
Order Now
Specificaties
Environmentally Preferable