Zwavelzuur 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

30144.294EA 31 EUR
30144.294 30144.328 30144.363 30144.408
Zwavelzuur 0.5 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Zwavelzuur
Formule: H₂SO₄
Dichtheid: 1,01 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR