SCV gas sampling bags, Tedlar®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30274-U 30244-U 30272-U 30273-U 30276-U 30285-U
30274-UEA 262 EUR
30274-U 30244-U 30272-U 30273-U 30276-U 30285-U
SCV gas sampling bags, Tedlar®
Zakken Gaszakken
These screw cap valve (SCV) bags are suitable for calibration mixes - gas phase standards/gas mixtures.