Sodium periodate ≥99.8%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

30323-100G 30323-500G 30323-6X500G
30323-100GEA 98.9 EUR
30323-100G 30323-500G 30323-6X500G
Sodium periodate ≥99.8%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium periodate
Formule: NaIO₄
Dichtheid: 3,86 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003534
CAS nummer: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1,I
Merck Index: 13,08714

Order Now

Specification Test Results

Assay Min. 99.8 %
Assay (excluding dried substances) 99.8 - 100.3 %
Loss on drying (110°C) Max. 0.2 %
Iron (Fe) Max. 0.001 %
Manganese (Mn) Max. 0.0001 %
Heavy metals (as Pb) Max. 0.001 %
Other halogens (as Cl) Max. 0.01 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR