Sodium hypophosphite monohydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium phosphinate monohydrate

30410-50KG 30410-1KG
30410-50KGEA 3610 EUR
30410-50KG 30410-1KG
Sodium hypophosphite monohydrate ≥99%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Sodium hypophosphite monohydrate
Formule: NaH₂PO₂·H₂O
MW: 105.99 g/mol
Smeltpunt: 90
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06252379
CAS nummer: 10039-56-2
EINECS: 231-669-9

Order Now

Specification Test Results

Assay Min. 99 %
pH (5 %, 20°C) 6-8
Ammonium (NH₄) Max. 0.001 %
Arsenic (As) Max. 0.0001 %
Barium (Ba) Max. 0.005 %
Calcium (Ca) Max. 0.02 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Magnesium (Mg) Max. 0.005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Chloride (Cl) Max. 0.005 %
Phosphate, phosphite (as PO₄) Max. 0.5 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR