Basacryl Red GL 19% (dye content), Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Basic Red 29

306525-25G
306525-25GEA 128 EUR
306525-25G
Basacryl Red GL 19% (dye content), Sigma-Aldrich®
Basacryl Red GL

Basacryl Red GL, Dye content 19 % (Basic Red 29)

Formule: C₁₉H₁₇ClN₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013337
CAS nummer: 42373-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=809da5cfba5f93a391f67186c543b6e95f501166&vid=a4bde9eff209ed4ab902572b1e42c12b0d3847cd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR