Zwavelzuur 95,0-97,0, Puriss. p.a. Reag. Ph. Eur., ACS, ISO Hg_determination, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

30743-50KG 30743-1L 30743-2.5L 30743-2.5L-GL 30743-4X2.5L 30743-4X2.5LGL 30743-4X5L 30743-5L 30743-6X1L 30743-4X2.5L-G 30743-1L-GL 30743-185L
30743-50KGEA 412 EUR
30743-50KG 30743-1L 30743-2.5L 30743-2.5L-GL 30743-4X2.5L 30743-4X2.5LGL 30743-4X5L 30743-5L 30743-6X1L 30743-4X2.5L-GL 30743-1L-GL HONC30743-185L
Zwavelzuur 95,0-97,0, Puriss. p.a. Reag. Ph. Eur., ACS, ISO Hg_determination, Fluka™
Zwavelzuur
Formule: H₂SO₄
MW: 98.08 g/mol
Kookpunt: 310 °C (1.013 hPa)
Smeltpunt: –14…–10 °C
Dichtheid: 1.840 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Order Now

Specification Test Results

Vapor Pressure 1 mmHg ( 146 °C)
Assay 95.0 - 97.0 %
Residue on ignition Max. 5 ppm
Ammonium (NH₄) Max. 2 ppm
Silver (Ag) Max. 0.000002 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.000005 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.000005 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000002 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000005 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.1 ppm
Germanium (Ge) Max. 0.000005 %
Mercury (Hg) Max. 0.005 ppm
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.000005 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000002 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Chloride (Cl) Max. 0.1 ppm
Nitrate (NO₃) Max. 0.2 ppm
Phosphate (PO₄) Max. 0.00005 %
KMnO₄ reducing matter (as O) Max. 0.0002 %
APHA Max. 10
KMnO₄ reducing matter (as SO₂) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR