Tetramethylammonium perchlorate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

30834.14 30834.22
30834.14EA 134 EUR
30834.14 30834.22
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate

CAS No.: 2537-36-2 Molecular Formula: (CH3)4NClO4 Formula Weight: 173.60 Physical Form: Crystalline Melting Point: 300° UN No.: 1479 MDL No.: MFCD00043207

Formule: C₄H₁₂ClNO₄
Smeltpunt: >300 °C
Dichtheid: 1,379 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043207
CAS nummer: 2537-36-2
EINECS: 219-805-5
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR