Natriumthiosufaat 0.01 mol/l (0.01 N) in aqueous solution, VOLUSOL® volumetric solution

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

309337.1000EA 39 EUR
309337.1000
Natriumthiosufaat 0.01 mol/l (0.01 N) in aqueous solution, VOLUSOL® volumetric solution
Natriumthiosufaat
Formule: Na₂S₂O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.00998 - 0.01002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR