Broad range RNA quantitation kits, AccuBlue®
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31073-T 31073
31073-TEA 146 EUR
31073-T 31073.
Broad range RNA quantitation kits, AccuBlue®
Testen Nucleïnezuurbepalingen
The AccuBlue® broad range RNA quantitation kit boasts the widest linear range (5 to 1000 ng RNA) of available RNA quantitation kits, while providing exceptional accuracy, sensitivity, and high RNA selectivity.
  • Detection range spans Qubit® HS and BR
  • Highly selective for RNA over dsDNA
  • Far-red fluorescence emission
  • For fluorescence microplate reader or Qubit® RNA broad range program
  • Mammalian RNA standard to accurately quantify similar samples
High sensitivity and broad linear range:
The assay is linear between 5 and 1000 ng of RNA per 200 μl assay well, which spans both Qubit® high sensitivity and Qubit® broad range detection ranges. It is ideal for use in quantifying RNA for sensitive applications such as Next-Gen Sequencing (NGS) or reverse transcription PCR (RT-PCR).
RNA specificity:
Unlike absorbance-based measurements, RNA broad range dye is selective for RNA over double-stranded DNA, and can tolerate an equimolar amount of dsDNA in the sample without significant effect on RNA quantitation (purified RNA samples are still recommended).
Instrument compatibility:
The AccuBlue® broad range RNA quantitation assay is designed for use with fluorescence microplate readers equipped with excitation and emission filters for detecting far-red fluorescence. It is also optimised for use in the Qubit® fluorometer, using the pre-programmed RNA Broad Range program.
Bestelinformatie: Replacement vials of RNA standard can be purchased separately.
Leveringsinformatie: Kit components: RNA broad range dye 200X, RNA broad range buffer, RNA dilution buffer (Standard 1 if using Qubit®), RNA broad range standard, 100 ng/μl (Standard 2 if using Qubit®).
Order Now
Specificaties
Environmentally Preferable