Potassium sulphate ≥99%, Puriss. p.a. ACS, ISO, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

31270-1KG 31270-5KG 31270-6X1KG
31270-1KGEA 74.5 EUR
31270-1KG 31270-5KG 31270-6X1KG
Potassium sulphate ≥99%, Puriss. p.a. ACS, ISO, Fluka™
Potassium sulphate
Formule: K₂SO₄
MW: 174.26 g/mol
Kookpunt: 1.698 °C (1.013 hPa)
Smeltpunt: 1.067 °C
Dichtheid: 2.66 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011388
CAS nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay Min. 99.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 5.5 - 8.0
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 5 ppm
Magnesium (Mg) Max. 0.002 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.0005 %
Total N Max. 5 ppm
Phosphate (PO₄) Max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR