1,5-Diphenylcarbazide ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

31390.06 31390.14 31390.22
31390.06EA 29.6 EUR
31390.06 31390.14 31390.22
1,5-Diphenylcarbazide ACS
1,5-Diphenylcarbazide

CAS No.: 140-22-7 Molecular Formula: C13H14N4O Formula Weight: 242.28 Physical Form: Crystalline Sensitivity: Air and Light Sensitive Appearance: White to cream Melting Point: 170-176° Solubility: Soluble in hot alcohol, acetone, glacial acetic acid Beilstein Reference: 752039 Merck Reference: 14,3321 MDL No.: MFCD00003013

Formule: C₁₃H₁₄N₄O
Kookpunt: 454 °C (1013 hPa)
Smeltpunt: 170…176 °C
Dichtheid: 1,31 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003013
CAS nummer: 140-22-7
EINECS: 205-403-7
Merck Index: 14,3321
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR