Kaliumhydroxide, pellets NF, Multi-Compendial, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

3152-31EA 84.2 EUR
3152-31 3152-35 3152-37
Kaliumhydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Kaliumhydroxide
Formule: KOH
Kookpunt: 1320 °C (1013 hPa)
Smeltpunt: 360 °C
Dichtheid: 2,04 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
Bereik: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Insoluble Substances Passes Test
NF - Content of Potassium ≥59.9 %
NF - Limit of Sodium ≤1.0 %
NF - Assay (Total Alkali) ≥85.0 %
NF - Carbonate (as K₂CO₃) ≤3.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Carbonate (as K₂CO₃) ≤2.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Phosphate (PO₄) ≤100 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Sodium (Na) ≤1.0 %
EP/BP - Assay 85.0 - 100.5 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Sodium (Na) Passes Test
JP - Potassium Carbonate (K₂CO₃) ≤2.0 %
JP - Assay (KOH) ≥85.0 %
No Class 1,2,3 or other solvents are used or produced in the = 85.0 %
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR