Natriumthiosufaat 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

31547.293EA 25.9 EUR
31547.293
Natriumthiosufaat 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Natriumthiosufaat
Formule: Na₂S₂O₃
MW: 158.11 g/mol
Kookpunt: 100
Smeltpunt: 48
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR