Morfoline ≥99.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

31984.36 31984.57
31984.36EA 56.6 EUR
31984.36 31984.57
Morfoline ≥99.0% ACS
Morfoline

Purity: 99.0% min CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972

Formule: C₄H₉NO
Kookpunt: 129 °C
Smeltpunt: –5 °C
Dichtheid: 1,007
Vlampunt: 35 °C (95 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005972
CAS nummer: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
Merck Index: 14,6277
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR