Zwavelzuur concentrate for dilution to 0.5 mol aqueous solution, ConvoL NORMADOSE® Volumetric

Leverancier: VWR Chemicals
Ratings:
Avg. Ratings: 5.0 (1 Review)

Total Ratings: 1
Avg. Ratings: out of 5

Gevaar


conc_1l

32053.602EA 25.7 EUR
32053.602
Zwavelzuur concentrate for dilution to 0.5 mol aqueous solution, ConvoL NORMADOSE® Volumetric
Zwavelzuur
Formule: H₂SO₄
MW: 98.08 g/mol
Kookpunt: 330
Smeltpunt: 10.38
Dichtheid: 1.84
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Titer (20°C) 0.4975 - 0.5025 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR