Kaliumdichromaat concentrate for dilution to 1/60 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Dose for preparation of 1 litre of solution.

32061.600EA 34.4 EUR
32061.600
Kaliumdichromaat concentrate for dilution to 1/60 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Kaliumdichromaat
Formule: K₂Cr₂O₇
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011367
CAS nummer: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Titer (20°C) 0.01658 - 0.01675 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR