4'-Hydroxydiclofenac, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

32412-10MG
32412-10MGEA 585 EUR
32412-10MG
4'-Hydroxydiclofenac, Supelco®
4'-Hydroxydiclofenac
Formule: C₁₄H₁₁Cl₂NO₃
MW: 312.15 g/mol
MDL Nummer: MFCD01671980
CAS nummer: 64118-84-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37afae48126ea811d017602749c7899ad1eff7d6&vid=e370be4d61351969b22671ea4cc4cdb0ce2e9de3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR