Tetrachloorethyleen 99+%, Ultrapure for spectrophotometry

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

32437.K2 32437.K7 32437.M1
32437.K2EA 114 EUR
32437.K2 32437.K7 32437.M1
Tetrachloorethyleen 99+%, Ultrapure for spectrophotometry
Tetrachloorethyleen

Purity: 99+% CAS No.: 127-18-4 Molecular Formula: Cl2C=CCl2 Formula Weight: 165.83 Physical Form: Liquid Density: 1.623 Refractive Index: 1.5055 Odour: Ether-like Melting Point: -22° Boiling Point: 120-121° Beilstein Reference: 1361721 Merck Reference: 14,9190 UN No.: 1897 MDL No.: MFCD00000834

Formule: C₂Cl₄
Kookpunt: 120…121 °C
Smeltpunt: –22 °C
Dichtheid: 1,623
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000834
CAS nummer: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1,III
Merck Index: 14,9190
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR