Lithium tetrahydroborate ≥95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

32563.09 32563.18
32563.09EA 160 EUR
32563.09 32563.18
Lithium tetrahydroborate ≥95%
Lithium tetrahydroborate

Purity: 95% CAS No.: 16949-15-8 Molecular Formula: BH4Li Formula Weight: 21.78 Physical Form: Powder Density: 0.66 Appearance: White Odour: Odourless Melting Point: ca 280° dec. Solubility: Soluble in ether, THF, and aliphatic amines Merck Reference: 14,5525 UN No.: 1413 MDL No.: MFCD00011088


Strong reducing agent. Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides

Formule: H₄BLi
Kookpunt: 66 °C (1013 hPa)
Dichtheid: 0,66
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011088
CAS nummer: 16949-15-8
EINECS: 241-021-7
UN: 1413
ADR: 4.3,I
Merck Index: 14,5525

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR