Cyclohexaan, extra dry 99.5%, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

326831000 326830010 326830025
326831000.EA 27.4 EUR
326831000. 326830010. 326830025.
Cyclohexaan, extra dry 99.5%, AcroSeal®
Cyclohexaan
Formule: C₆H₁₂
Kookpunt: 81 °C (760 mmHg)
Dichtheid: 0,77 g/cm³
Vlampunt: –18 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003814
CAS nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.4 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.001 meq/g

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR