2,2,4-Trimethylpentane, dry_extra 99.5%, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

326941000 326940010
326941000.EA 28.8 EUR
326941000. 326940010.
2,2,4-Trimethylpentane, dry_extra 99.5%, AcroSeal®
2,2,4-Trimethylpentane
Formule: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Kookpunt: 98…99 °C (760 mmHg)
Smeltpunt: –107 °C
Dichtheid: 0.69 g/cm³
Vlampunt: –12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.4 %
Residue after evaporation ≤0.0005 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR