Malachietgroen oxalaat in water, crystalline for microscopy, Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Malachite green hemioxalate, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate, Basic Green 4, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)

M-Clarity™ quality level = MQ200

32745-100G 32745-1KG 32745-50G
32745-100GEA 55.9 EUR
32745-100G 32745-1KG 32745-50G
Malachietgroen oxalaat in water, crystalline for microscopy, Sigma-Aldrich®
Malachietgroen oxalaat
Formule: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
Smeltpunt: ∼159 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011766
CAS nummer: 2437-29-8
EINECS: 219-441-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,05722
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2dffa05d1d66e7e32371284ada0fcf1a0faa7d0&vid=950afba5ed94b79530cd91b1261e3c2223d2aecd&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7723ac34e7273345d2b069c0836679df388eaaf7&vid=ec3b640e950ddc753e47d460407fdd3277490d06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.01 g in 10 ml methanol
Loss on drying ≤7.0%
Wavelength (1) UV 609 - 619 nm
Wavelength (2) UV 420 - 430 nm
Extinction (1%/1 cm) (1) (UV) ≥300


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR