Mierenzuur 100% ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

33015-1L 33015-2.5L 33015-500ML
33015-1LEA 64.4 EUR
33015-1L 33015-2.5L 33015-500ML
Mierenzuur 100% ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Mierenzuur
Formule: HCO₂H
Kookpunt: 100,4 °C (1.013 hPa)
Smeltpunt: 4 °C
Dichtheid: 1,22 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003297
CAS nummer: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Density (D 20/20) 1.217 - 1.223
Non-volatile matter Max. 0.001 %
Ammonium (NH₄) Max. 0.001 %
Silver (Ag) Max. 0.000002 %
Aluminium (Al) Max. 0.000005 %
Barium (Ba) Max. 0.000005 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000005 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.0002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000002 %
Magnesium (Mg) Max. 0.00005 %
Manganese (Mn) Max. 0.000005 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000005 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000002 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000005 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.0005 %
Oxalate (C₂O₄) Max. 0.005 %
Sulfate (SO₄) Max. 0.0005 %
Sulfite (SO₃) complying
Acetic acid (CH₃COOH) Max. 0.005 %
APHA Max. 10
Solubility in water complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR