Detectable spoons, curved, long handle, blue, SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5378-5012
331-0338EA 31.3 EUR
331-0338
Detectable spoons, curved, long handle, blue, SteriPlast®
Lepels
The spoon with curved spoon bowl is suitable for a wide range of different media including powders, granulates, pastes and liquids. The angled spoon shape is also good for reaching into the corners of a container. The long handle of the plastic spoon makes it possible to empty residue from large containers easily and can also reach into places that are hard to reach such as jerrycans with narrow necks.
  • PS detectable
  • Individually packaged and sterilised, for single use
  • With curved spoon form, handle length 300 mm
  • Can be detected by colour scanners, metal
More Product Information